missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde 97.0+%, TCI America™
Supplier: TCI America M240425G
Specifications
| 2-Methyl-3-(3,4-methylenedioxyphenyl)propionaldehyde | |
| Yellow | |
| C11H12O3 | |
| 25 g | |
| BOPPSUHPZARXTH-UHFFFAOYNA-N | |
| 3-(2H-1,3-benzodioxol-5-yl)-2-methylpropanal | |
| 64805 | |
| ≥97.0% (GC) |
| 1205-17-0 | |
| 154°C | |
| MFCD00067053 | |
| 2-methyl-3-3,4-methylenedioxyphenyl propanal, 3-1,3-benzodioxol-5-yl-2-methylpropanal, alpha-methyl-3,4-methylene-dioxyhydrocinnamic aldehyde, 1,3-benzodioxole-5-propanal, alpha-methyl, 1,3-benzodioxole-5-propanal, .alpha.-methyl, 3-3,4-methylenedioxyphenyl-2-methylpropanal, alpha-methyl-1,3-benzodioxole-5-propionaldehyde, 2-methyl-3-3,4-methylenedioxyphenyl-propanal, 3-3,4-methylenedioxyphenyl-2-methyl-propanol, 2-methyl-3-3,4-methylenedioxyphenyl propionaldehyde | |
| CC(CC1=CC=C2OCOC2=C1)C=O | |
| 192.21 | |
| 192.21 | |
| Liquid |
Chemical Identifiers
| 1205-17-0 | |
| 192.21 | |
| BOPPSUHPZARXTH-UHFFFAOYNA-N | |
| 64805 | |
| CC(CC1=CC=C2OCOC2=C1)C=O |
| C11H12O3 | |
| MFCD00067053 | |
| 2-methyl-3-3,4-methylenedioxyphenyl propanal, 3-1,3-benzodioxol-5-yl-2-methylpropanal, alpha-methyl-3,4-methylene-dioxyhydrocinnamic aldehyde, 1,3-benzodioxole-5-propanal, alpha-methyl, 1,3-benzodioxole-5-propanal, .alpha.-methyl, 3-3,4-methylenedioxyphenyl-2-methylpropanal, alpha-methyl-1,3-benzodioxole-5-propionaldehyde, 2-methyl-3-3,4-methylenedioxyphenyl-propanal, 3-3,4-methylenedioxyphenyl-2-methyl-propanol, 2-methyl-3-3,4-methylenedioxyphenyl propionaldehyde | |
| 3-(2H-1,3-benzodioxol-5-yl)-2-methylpropanal |
Safety and Handling
EINECSNumber : (5)-3560
RTECSNumber : DF4922185
TSCA : Yes