missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2h-indazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC44401CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC44401CB 250 mg
CC44401DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC44401CB Supplier Thermo Scientific Chemicals Supplier No. CC44401CB

Chemical Identifiers

CAS 34252-44-3
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
InChI Key ABRISPFTOZIXMP-UHFFFAOYSA-N
Synonym 2-methyl-2h-indazole-3-carboxylic acid, 2h-indazole-3-carboxylic acid, 2-methyl
PubChem CID 7060537
IUPAC Name 2-methylindazole-3-carboxylic acid
SMILES CN1C(=C2C=CC=CC2=N1)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-2H-indazole-3-carboxylic acid