missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-thiopseudourea hemisulfate, 98%

Catalog No. p-168249
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127891000 100 g Plastic bottle
AC127895000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127891000 Supplier Thermo Scientific Chemicals Supplier No. 127891000

CAS: 867-44-7 | C2H6N21/2H2SO4 | 139.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 867-44-7
MDL Number MFCD00013055
InChI Key BZZXQZOBAUXLHZ-UHFFFAOYSA-N
Synonym s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
PubChem CID 13347
IUPAC Name methyl carbamimidothioate;sulfuric acid
SMILES CSC(=N)N.CSC(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 240.0°C to 241.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C2H6N21/2H2SO4
Linear Formula [CH3SC(=NH)NH2]2·1/2H2SO4
Quantity 100 g
Solubility Information Solubility in water: 260g/L (20°C). Other solubilities: 4g/L methanol (20°C)
Molecular Weight (g/mol) 139.18
Formula Weight 139.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Methyl-2-thiopseudourea hemisulfate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-759-7
RTECSNumber UM9200000
TSCA TSCA

RUO – Research Use Only