missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7126418
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076625G 25 g
D0766500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D076625G Supplier TCI America Supplier No. D076625G
Only null left

Chemical Identifiers

CAS 78-26-2
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.20
MDL Number MFCD00004696
InChI Key JVZZUPJFERSVRN-UHFFFAOYSA-N
Synonym 2-methyl-2-propyl-1,3-propanediol, 2,2-bis hydroxymethyl pentane, 1,3-propanediol, 2-methyl-2-propyl, 2-methyl-2-n-propyl-1,3-propanediol, unii-ylm5kru0p4, 2,2-dimethylolpentane, ylm5kru0p4, 2-methyl-2-propyl-propane-1,3-diol, carisoprodol impurity c, acmc-1basp
PubChem CID 66220
IUPAC Name 2-methyl-2-propylpropane-1,3-diol
SMILES CCCC(C)(CO)CO

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 132.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2-propyl-1,3-propanediol
EINECSNumber (2)-0240
RTECSNumber TY9386000
TSCA Yes