Learn More
2-Methyl-2-propen-1-ol, 98%
CAS: 513-42-8 | C4H8O | 72.11 g/mol
$95.68 - $95.68
Chemical Identifiers
| CAS | 513-42-8 |
|---|---|
| Molecular Formula | C4H8O |
| MDL Number | MFCD00004737 |
| InChI Key | BYDRTKVGBRTTIT-UHFFFAOYSA-N |
| PubChem CID | 10557 |
| IUPAC Name | 2-methylprop-2-en-1-ol |
| SMILES | CC(=C)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6050206
|
Thermo Scientific Chemicals
H6050206 |
5 g |
Each for $95.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 513-42-8 | |
| MFCD00004737 | |
| 10557 | |
| CC(=C)CO |
| C4H8O | |
| BYDRTKVGBRTTIT-UHFFFAOYSA-N | |
| 2-methylprop-2-en-1-ol |
Specifications
| 513-42-8 | |
| 113°C to 115°C | |
| C4H8O | |
| CH2=C(CH3)CH2OH | |
| 5 g | |
| 969226 | |
| BYDRTKVGBRTTIT-UHFFFAOYSA-N | |
| 2-methylprop-2-en-1-ol | |
| 10557 | |
| 98% |
| 0.857 | |
| 32°C (90°F) | |
| 1.426 | |
| MFCD00004737 | |
| UN2614 | |
| methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol | |
| CC(=C)CO | |
| 72.107 | |
| 72.11 | |
| 2-Methyl-2-propen-1-ol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHALLYL ALCOHOL
EINECSNumber : 208-161-0
RTECSNumber : UD5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only