missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™

Catalog No. p-4323268
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438751000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438751000 Supplier Thermo Scientific Chemicals Supplier No. 438751000

CAS: 35293-35-7 | C10H13ClMg | 192.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35293-35-7 , 60-29-7
Molecular Formula C10H13ClMg
Molecular Weight (g/mol) 192.97
MDL Number MFCD00075586
InChI Key BXKYFONHSSFLKR-UHFFFAOYSA-M
Synonym neophylmagnesium chloride, chloro 2-methyl-2-phenylpropyl magnesium, magnesium, chloro 2-methyl-2-phenylpropyl, 2-methyl-2-phenylpropylmagnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride solution, neophyl magnesium chloride, bxkyfonhssflkr-uhfffaoysa-m, 2-methyl-2-phenylpropyl magnesium chloride, 2-methyl-2-phenyl-propanyl magnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride solution, 0.5 m in diethyl ether
PubChem CID 118802
IUPAC Name magnesium;2-methanidylpropan-2-ylbenzene;chloride
SMILES CC(C)(C[Mg]Cl)C1=CC=CC=C1

Specifications

CAS Min % 10.16, 84.77
CAS Max % 15.23, 89.84
Color Colorless to Yellow
Density 0.7600g/mL
Flash Point −28°C
Packaging AcroSeal™ Glass Bottle
Linear Formula C6H5C(CH3)2CH2MgCl
Quantity 100 mL
Specific Gravity 0.76
Formula Weight 192.97
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-phenylpropylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any
EINECSNumber 252-492-3

RUO – Research Use Only