missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-pentene 95.0+%, TCI America™
Supplier: TCI America M054125ML
Specifications
| 2-Methyl-2-pentene | |
| Yellow | |
| C6H12 | |
| 25 mL | |
| 2-methyl-2-pentene, 2-pentene, 2-methyl, 4-methyl-3-pentene, pentene, methyl, unii-7kog6sh4s6, 7kog6sh4s6, butene, dimethyl, 2-methyl-pentene-2, 1,1-dimethyl-1-butene, trans-2-methyl-2-pentene | |
| CCC=C(C)C | |
| 84.162 | |
| 84.16 | |
| Liquid |
| 625-27-4 | |
| 67°C | |
| MFCD00009382 | |
| 2288 | |
| JMMZCWZIJXAGKW-UHFFFAOYSA-N | |
| 2-methylpent-2-ene | |
| 12243 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 625-27-4 | |
| 84.162 | |
| JMMZCWZIJXAGKW-UHFFFAOYSA-N | |
| 12243 | |
| CCC=C(C)C |
| C6H12 | |
| MFCD00009382 | |
| 2-methyl-2-pentene, 2-pentene, 2-methyl, 4-methyl-3-pentene, pentene, methyl, unii-7kog6sh4s6, 7kog6sh4s6, butene, dimethyl, 2-methyl-pentene-2, 1,1-dimethyl-1-butene, trans-2-methyl-2-pentene | |
| 2-methylpent-2-ene |
Safety and Handling
EINECSNumber : (2)-0031
RTECSNumber : SB2240000
TSCA : Yes