missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentene 95.0+%, TCI America™

Catalog No. M05415ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05415ML 5 mL
M054125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05415ML Supplier TCI America Supplier No. M05415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-27-4
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009382
InChI Key JMMZCWZIJXAGKW-UHFFFAOYSA-N
Synonym 2-methyl-2-pentene, 2-pentene, 2-methyl, 4-methyl-3-pentene, pentene, methyl, unii-7kog6sh4s6, 7kog6sh4s6, butene, dimethyl, 2-methyl-pentene-2, 1,1-dimethyl-1-butene, trans-2-methyl-2-pentene
PubChem CID 12243
IUPAC Name 2-methylpent-2-ene
SMILES CCC=C(C)C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 mL
UN Number 2288
Formula Weight 84.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-pentene
EINECSNumber (2)-0031
RTECSNumber SB2240000
TSCA Yes