missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-pentanol 96.0+%, TCI America™
$63.38 - $249.78
Chemical Identifiers
| CAS | 590-36-3 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.18 |
| MDL Number | MFCD00004485 |
| InChI Key | WFRBDWRZVBPBDO-UHFFFAOYSA-N |
| Synonym | 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l |
| PubChem CID | 11543 |
| IUPAC Name | 2-methylpentan-2-ol |
| SMILES | CCCC(C)(C)O |
Chemical Identifiers
| 590-36-3 | |
| 102.18 | |
| WFRBDWRZVBPBDO-UHFFFAOYSA-N | |
| 11543 | |
| CCCC(C)(C)O |
| C6H14O | |
| MFCD00004485 | |
| 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l | |
| 2-methylpentan-2-ol |
Specifications
| 590-36-3 | |
| 122°C | |
| MFCD00004485 | |
| 2560 | |
| WFRBDWRZVBPBDO-UHFFFAOYSA-N | |
| 2-methylpentan-2-ol | |
| 11543 | |
| ≥96.0% (GC) | |
| 2-Methyl-2-pentanol |
| Yellow | |
| C6H14O | |
| 5 mL | |
| 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l | |
| CCCC(C)(C)O | |
| 102.18 | |
| 102.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes