missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentanol, 99%

Catalog No. AAA1909022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1909022 100 g
AAA1909014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1909022 Supplier Thermo Scientific Chemicals Supplier No. A1909022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-36-3 | C6H14O | 102.18 g/mol

It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

Solubility
Soluble in water at 20°C 32g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 590-36-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004485
InChI Key WFRBDWRZVBPBDO-UHFFFAOYSA-N
Synonym 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l
PubChem CID 11543
IUPAC Name 2-methylpentan-2-ol
SMILES CCCC(C)(C)O

Specifications

Melting Point -103°C
Density 0.81
Boiling Point 120°C to 122°C
Flash Point 30°C (86°F)
Odor Alcohol-like
Refractive Index 1.41
Quantity 100 g
UN Number UN2560
Beilstein 1718951
Solubility Information Soluble in water at 20°C 32g/L. Soluble in alcohol.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentanol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-METHYLPENTAN-2-OL
EINECSNumber 209-681-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only