missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-oxazoline 98.0+%, TCI America™

Catalog No. p-7132410
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
M085725G 25 g
M0857500G 500 g
2 options

Catalog No. M085725G

Supplier: TCI America M085725G

Only null left

Chemical Identifiers

CAS 1120-64-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005298
InChI Key GUXJXWKCUUWCLX-UHFFFAOYSA-N
Synonym 2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
PubChem CID 70713
ChEBI CHEBI:53614
IUPAC Name 2-methyl-4,5-dihydro-1,3-oxazole
SMILES CC1=NCCO1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
UN Number 1993
Formula Weight 85.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-oxazoline