missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-hexene 97.0+%, TCI America™

Catalog No. M05261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05261G 1 g
M05265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05261G Supplier TCI America Supplier No. M05261G
Only null left

Chemical Identifiers

CAS 2738-19-4
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00027232
InChI Key BWEKDYGHDCHWEN-UHFFFAOYSA-N
PubChem CID 17656
IUPAC Name 2-methylhex-2-ene
SMILES CCCC=C(C)C

Specifications

Color Colorless
Boiling Point 95°C
Quantity 1 g
UN Number 3295
Formula Weight 98.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-hexene