missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-cyclopenten-1-one, 97%

Catalog No. AC270360050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270360050 5 g Glass bottle
AC270360010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270360050 Supplier Thermo Scientific Chemicals Supplier No. 270360050

CAS: 1120-73-6 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1120-73-6
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00012275
InChI Key ZSBWUNDRDHVNJL-UHFFFAOYSA-N
Synonym 2-methylcyclopent-2-enone, 2-methyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2-methyl, 2-methyl-2-cyclopentenone, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone, acmc-20af73, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl
PubChem CID 14266
IUPAC Name 2-methylcyclopent-2-en-1-one
SMILES CC1=CCCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9790g/mL
Boiling Point 158.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800
Linear Formula CH3C5H5(0)
Quantity 5 g
Beilstein 07, 52
Specific Gravity 0.979
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-cyclopenten-1-one, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai
TSCA TSCA

RUO – Research Use Only