missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butanol, 99%, pure

CAS: 75-85-4 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals 166620025

 View more versions of this product

Catalog No. AC166620025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2-butanol
75-85-4
Colorless
102.0°C
Authentic
Glass Bottle
1.4040 to 1.4060
MFCD00004478
01,388
15,7253
Solubility in water: 70.7g/L (25°C). Other solubilities: miscible with alcohol,ether,benzene,chloroform,,glycerol,oils and acetone
CCC(C)(C)O
88.15
3.7 mPa.s (25°C)
99%
Liquid
99%
-12.0°C
0.8000g/mL
20°C
98.5% min. (GC)
C5H12O
C2H5C(CH3)2OH
2.5 L
0.8
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
MSXVEPNJUHWQHW-UHFFFAOYSA-N
2-methylbutan-2-ol
6405
88.15
Pure

Chemical Identifiers

75-85-4
88.15
MSXVEPNJUHWQHW-UHFFFAOYSA-N
6405
CCC(C)(C)O
C5H12O
MFCD00004478
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
2-methylbutan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful in contact with skin.
May cause drowsiness or d

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Danger

EINECSNumber : 200-908-9

RTECSNumber : SC0175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only