Learn More
2-Methyl-2-butanol, 98%
CAS: 75-85-4 | C5H12O | 88.15 g/mol
$389.97 - $966.85
Chemical Identifiers
CAS | 75-85-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00004478 |
InChI Key | MSXVEPNJUHWQHW-UHFFFAOYSA-N |
Synonym | 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol |
PubChem CID | 6405 |
IUPAC Name | 2-methylbutan-2-ol |
SMILES | CCC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA183040F
|
Thermo Scientific Chemicals
A183040F |
2500 mL |
Each for $389.97
|
|
||||||
AAA183040D
|
Thermo Scientific Chemicals
A183040D |
10,000 mL |
Each for $966.85
|
|
||||||
Description
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
75-85-4 | |
88.15 | |
MSXVEPNJUHWQHW-UHFFFAOYSA-N | |
6405 | |
CCC(C)(C)O |
C5H12O | |
MFCD00004478 | |
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol | |
2-methylbutan-2-ol |
Specifications
75-85-4 | |
0.805 g/mL | |
20°C (68°F) | |
C5H12O | |
MFCD00004478 | |
UN1105 | |
14,7140 | |
Miscible with alcohol,ether,benzene,chloroform,glycerol,oils and acetone. | |
CCC(C)(C)O | |
88.15 | |
88.15 | |
Liquid |
-12°C | |
101°C to 103°C | |
0.98 | |
1.405 | |
2500 mL | |
1361351 | |
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol | |
MSXVEPNJUHWQHW-UHFFFAOYSA-N | |
2-methylbutan-2-ol | |
6405 | |
98% | |
2-Methyl-2-butanol |
Safety and Handling
GHS H Statement
H225-H332-H315-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
H225-H312+H332-H315-H318-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANOLS
EINECSNumber : 200-908-9
RTECSNumber : SC0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only