missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2,4-pentanediol, 99+%

Catalog No. AC219450500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219450500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219450500 Supplier Thermo Scientific Chemicals Supplier No. 219450500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107-41-5 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key SVTBMSDMJJWYQN-UHFFFAOYSA-N
Synonym hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
PubChem CID 7870
ChEBI CHEBI:62995
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(CC(C)(C)O)O

Specifications

Melting Point -40.0°C
Density 0.9220g/mL
Boiling Point 197.0°C
Flash Point 93°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4260 to 1.4280
Linear Formula CH3CH(OH)CH2C(CH3)2OH
Quantity 50 g
Beilstein 01,486
Fieser 01,693
Merck Index 15,4746
Specific Gravity 0.922
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and lower aliphathic,hydrocarbons
Viscosity 36 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99+%
Chemical Name or Material 2-Methyl-2, 4-pentanediol
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 203-489-

RUO – Research Use Only