missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2,4-pentanediol, 99%

CAS: 107-41-5 | C6H14O2 | 118.18 g/mol

Supplier:  Thermo Scientific Chemicals 150340025

 View more versions of this product

Catalog No. AC150340025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2, 4-pentanediol
98.5
-40.0°C
197.0°C
Authentic
Glass Bottle
1.4260 to 1.4280
MFCD00004547
01, 486
0.922
hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
SVTBMSDMJJWYQN-UHFFFAOYSA-N
2-methylpentane-2,4-diol
7870
CHEBI:62995
99%
107-41-5
100.0
0.9220g/mL
93°C
98.5% min. (GC)
C6H14O2
CH3CH(OH)CH2C(CH3)2OH
2.5 L
01,693
15, 4746
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and lower aliphathic,hydrocarbons
CC(CC(C)(C)O)O
118.18
36 mPa.s (20°C)
118.18
Slightly Viscous Liquid

Chemical Identifiers

107-41-5
118.18
SVTBMSDMJJWYQN-UHFFFAOYSA-N
7870
2-methylpentane-2,4-diol
C6H14O2
MFCD00004547
hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
CHEBI:62995
CC(CC(C)(C)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-489-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only