missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-(4-methylphenyl)morpholine, 97%

Catalog No. AAH5073203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5073203 1 g
AAH50732MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5073203 Supplier Thermo Scientific Chemicals Supplier No. H5073203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 902836-81-1 | C12H17NO | 191.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 902836-81-1
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD08060961
InChI Key NQYMBSUCKZVKFF-UHFFFAOYSA-N
Synonym 2-methyl-2-p-tolyl morpholine, 2-methyl-2-4-methylphenyl morpholine, 2-methyl-2-p-tolylmorpholine, 2-methyl-2-p-tolyl-morpholine, 2-4-methylphenyl-2-methylmorpholine, morpholine, 2-methyl-2-4-methylphenyl
PubChem CID 24208807
IUPAC Name 2-methyl-2-(4-methylphenyl)morpholine
SMILES CC1=CC=C(C=C1)C2(CNCCO2)C

Specifications

Melting Point 49°C to 51°C
Quantity 1 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-(4-methylphenyl)morpholine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only