missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1314216-34-6
Molecular Formula C14H19BN2O2
Molecular Weight (g/mol) 258.13
InChI Key CPQGLMCLTALRHF-UHFFFAOYSA-N
Synonym 2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
PubChem CID 46738035
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445160010
SDS View Documents
Thermo Scientific Chemicals
445160010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445160050
SDS View Documents
Thermo Scientific Chemicals
445160050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1314216-34-6
258.13
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
C14H19BN2O2
CPQGLMCLTALRHF-UHFFFAOYSA-N
46738035

Specifications

1314216-34-6
97%
C14H19BN2O2
2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
258.13
258.13
2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
Authentic
Glass Bottle
1g
CPQGLMCLTALRHF-UHFFFAOYSA-N
46738035
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only