missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%

Catalog No. AC445160010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445160010 1 g
AC445160050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445160010 Supplier Thermo Scientific Chemicals Supplier No. 445160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1314216-34-6 | C14H19BN2O2 | 258.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314216-34-6
Molecular Formula C14H19BN2O2
Molecular Weight (g/mol) 258.13
MDL Number MFCD13176536
InChI Key CPQGLMCLTALRHF-UHFFFAOYSA-N
Synonym 2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
PubChem CID 46738035
IUPAC Name 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
SMILES CC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 211°C to 212°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only