missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-naphthyl Acetate 98.0+%, TCI America™
$57.79 - $344.86
Chemical Identifiers
| CAS | 5697-02-9 |
|---|---|
| Molecular Formula | C13H12O2 |
| Molecular Weight (g/mol) | 200.24 |
| InChI Key | WVOAPRDRMLHUMI-UHFFFAOYSA-N |
| Synonym | Acetic Acid 2-Methyl-1-naphthyl Ester, 1-Acetoxy-2-methylnaphthalene |
| PubChem CID | 11805727 |
| IUPAC Name | (2-methylnaphthalen-1-yl) acetate |
| SMILES | CC1=C(C2=CC=CC=C2C=C1)OC(=O)C |
Chemical Identifiers
| 5697-02-9 | |
| 200.24 | |
| Acetic Acid 2-Methyl-1-naphthyl Ester, 1-Acetoxy-2-methylnaphthalene | |
| (2-methylnaphthalen-1-yl) acetate |
| C13H12O2 | |
| WVOAPRDRMLHUMI-UHFFFAOYSA-N | |
| 11805727 | |
| CC1=C(C2=CC=CC=C2C=C1)OC(=O)C |
Specifications
| 5697-02-9 | |
| C13H12O2 | |
| Acetic Acid 2-Methyl-1-naphthyl Ester, 1-Acetoxy-2-methylnaphthalene | |
| CC1=C(C2=CC=CC=C2C=C1)OC(=O)C | |
| 200.24 | |
| 200.24 | |
| Crystalline Powder |
| 82°C | |
| 25 g | |
| WVOAPRDRMLHUMI-UHFFFAOYSA-N | |
| (2-methylnaphthalen-1-yl) acetate | |
| 11805727 | |
| ≥98.0% (GC) | |
| 2-Methyl-1-naphthyl Acetate |
Safety and Handling
TSCA : No