missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-hexene 97.0+%, TCI America™

Catalog No. M05251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05251G 1 g
M05255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05251G Supplier TCI America Supplier No. M05251G
Only null left

Chemical Identifiers

CAS 6094-02-6
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00009486
InChI Key IRUDSQHLKGNCGF-UHFFFAOYSA-N
PubChem CID 22468
IUPAC Name 2-methylhex-1-ene
SMILES CCCCC(=C)C

Specifications

Color Colorless
Boiling Point 92°C
Quantity 1 g
UN Number 3295
Formula Weight 98.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-hexene