missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Methyl-1-butanol, 98%

CAS: 137-32-6 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals B218250F

 View more versions of this product

Catalog No. AAB218250F


Only null left
Explore available promotions

Description

Description

2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.

Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

(+/-)-2-Methyl-1-butanol
<-70°C
129°C to 131°C
C5H12O
MFCD00004743
UN1105
14,6030
Soluble in water. [183g/L (0 C)] and alcohol.
CCC(C)CO
88.15
CHEBI:48945
98%
137-32-6
0.815
43°C (109°F)
1.411
2500 mL
1718810
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
QPRQEDXDYOZYLA-UHFFFAOYSA-N
2-methylbutan-1-ol
8723
88.15

Chemical Identifiers

137-32-6
88.15
QPRQEDXDYOZYLA-UHFFFAOYSA-N
8723
2-methylbutan-1-ol
C5H12O
MFCD00004743
2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
CHEBI:48945
CCC(C)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H335
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

EINECSNumber : 205-289-9

RTECSNumber : EL5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only