missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,8-naphthyridine 98.0+%, TCI America™

Catalog No. M23521G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23521G Supplier TCI America Supplier No. M23521G

Chemical Identifiers

CAS 1569-16-0
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD04066717
InChI Key FSWRUYCICUXURT-UHFFFAOYSA-N
Synonym 2-methyl-1,8 naphthyridine, 2-methyl 1,8-naphthyridine, 2-methyl-1,8-naphthyridine, 1,8-naphthyridine, 2-methyl, 2-methylpyridino 2,3-b pyridine, 2-methyl-1,8-naphtyridine, 2-methyl 1,8 naphthyridine, methylnaphthyridine, 2-methylnaphthyridine
PubChem CID 74073
IUPAC Name 2-methyl-1,8-naphthyridine
SMILES CC1=NC2=C(C=CC=N2)C=C1

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,8-naphthyridine