missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,8-naphthyridine, 97%, Thermo Scientific Chemicals

Catalog No. p-7032533
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3403203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3403203 Supplier Thermo Scientific Chemicals Supplier No. H3403203

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1569-16-0
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD04066717
InChI Key FSWRUYCICUXURT-UHFFFAOYSA-N
Synonym 2-methyl-1,8 naphthyridine, 2-methyl 1,8-naphthyridine, 2-methyl-1,8-naphthyridine, 1,8-naphthyridine, 2-methyl, 2-methylpyridino 2,3-b pyridine, 2-methyl-1,8-naphtyridine, 2-methyl 1,8 naphthyridine, methylnaphthyridine, 2-methylnaphthyridine
PubChem CID 74073
IUPAC Name 2-methyl-1,8-naphthyridine
SMILES CC1=NC2=C(C=CC=N2)C=C1

Specifications

Melting Point 91°C to 93°C
Quantity 1 g
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-1,8-naphthyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only