missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-hexadiene 98.0+%, TCI America™

Catalog No. M03395ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03395ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03395ML Supplier TCI America Supplier No. M03395ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4049-81-4
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00008603
InChI Key SLQMKNPIYMOEGB-UHFFFAOYSA-N
Synonym 2-Methyldiallyl, sym-Vinylisopropenylethane
PubChem CID 19965
IUPAC Name 2-methylhexa-1,5-diene
SMILES CC(=C)CCC=C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,5-hexadiene