missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Catalog No. CC05713CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC05713CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC05713CB Supplier Thermo Scientific Chemicals Supplier No. CC05713CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103694-26-4
Molecular Formula C5H8N2S
Molecular Weight (g/mol) 128.193
MDL Number MFCD06212804
InChI Key ZCKAEFOHSOQKHN-UHFFFAOYSA-N
Synonym 2-methylthiazol-4-yl methanamine, 2-methyl-1,3-thiazole-4-methylamine, 2-methyl-1,3-thiazol-4-yl methanamine, 2-methyl-1,3-thiazol-4-yl methylamine, c-2-methyl-thiazol-4-yl-methylamine, 4-thiazolemethanamine, 2-methyl, 4-thiazolemethanamine,2-methyl, 4-aminomethyl-2-methyl-1,3-thiazole, 1-2-methyl-1,3-thiazol-4-yl methanamine, 2-methylthiazol-4-yl methylamine
PubChem CID 18467479
IUPAC Name (2-methyl-1,3-thiazol-4-yl)methanamine
SMILES CC1=NC(=CS1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 128.2
Percent Purity 97%
Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.