missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. M1114500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1114500ML 500 mL
M111425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1114500ML Supplier TCI America Supplier No. M1114500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2163-42-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00082586
InChI Key QWGRWMMWNDWRQN-UHFFFAOYSA-N
Synonym 1,3-Dihydroxy-2-methylpropane
PubChem CID 75103
IUPAC Name 2-methylpropane-1,3-diol
SMILES CC(CO)CO

Specifications

Color Yellow
Boiling Point 214°C
Quantity 500 mL
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-propanediol
EINECSNumber (2)-3203
TSCA Yes