missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Methyl-1,3-propanediol

Catalog No. 3757213L
Encompass
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3757213L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3757213L Supplier Sigma Aldrich Supplier No. 3757213L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methyl-1,3-propanediol

Specifications

CAS 2163-42-0
Melting Point -91°C
Density 1.015 g/mL (at 25°C)
Boiling Point 123°C to 125°C (20 mmHg)
Molecular Formula C4H10O2
Refractive Index n20/D 1.445 (literature)
Linear Formula CH3CH(CH2OH)2
Quantity 3 L
MDL Number MFCD00082586
Synonym 1,3-Dihydroxy-2-methylpropane; 2-Methylpropan-1,3-diol; Methylpropanediol
Molecular Weight (g/mol) 90.12
Percent Purity 99%
Show More Show Less