missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-propanediamine 98.0+%, TCI America™

Catalog No. M16575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16575G 5 g
M16571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16575G Supplier TCI America Supplier No. M16575G
Only null left

Chemical Identifiers

CAS 2400-78-4
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD08276399
InChI Key BDXGMDGYOIWKIF-UHFFFAOYSA-N
Synonym 1,3-Diamino-2-methylpropane
PubChem CID 14499052
IUPAC Name 2-methylpropane-1,3-diamine
SMILES CC(CN)CN

Specifications

Color Colorless
Quantity 5 g
UN Number 2734
Formula Weight 88.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1,3-propanediamine