missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-methyl-1,3-oxazole-4-carboxylic acid, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 127.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-methyl-1, 3-oxazole-4-carboxylic acid |