missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-1,3-oxazole-4-carboxylic acid, Thermo Scientific™

Catalog No. CC57901CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC57901CB 250mg Amber Glass Bottle
CC57901DA 1g Amber Glass Bottle
CC57901DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC57901CB Supplier Thermo Scientific Chemicals Supplier No. CC57901CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23062-17-1
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
InChI Key IARMCEYEYXXEOS-UHFFFAOYSA-N
Synonym 2-methyloxazole-4-carboxylic acid, 2-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 2-methyl, 4-carboxy-2-methyl-1,3-oxazole, 2-methyloxa, oxazole-4-carboxylic acid, 2-methyl, 2-methyloxazol-4-carboxylic acid, 4-oxazolecarboxylicacid, 2-methyl, 2-methyl-3h-isoxazole-4-carboxylic acid, 2-methyl-4-oxazolecarboxylic acid
PubChem CID 539404
IUPAC Name 2-methyl-1,3-oxazole-4-carboxylic acid
SMILES CC1=NC(=CO1)C(=O)O

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 2-methyl-1, 3-oxazole-4-carboxylic acid