missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-1,3-oxazole-4-carbaldehyde, Thermo Scientific™

Catalog No. p-4391412
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC57904CB 250mg Amber Glass Bottle
CC57904DA 1g Amber Glass Bottle
CC57904DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC57904CB Supplier Thermo Scientific Chemicals Supplier No. CC57904CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113732-84-6
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
InChI Key ARAUEWKXKTYCHZ-UHFFFAOYSA-N
Synonym 2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde
PubChem CID 11073372
IUPAC Name 2-methyl-1,3-oxazole-4-carbaldehyde
SMILES CC1=NC(=CO1)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-methyl-1, 3-oxazole-4-carbaldehyde