missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dithiane, 98%

CAS: 6007-26-7 | C5H10S2 | 134.255 g/mol

Supplier:  Thermo Scientific Chemicals L0020906

 View more versions of this product

Catalog No. AAL0020906

Only null left
Explore available promotions

Description

Description

It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Methyl-1,3-dithiane
1.212
82°C (179°F)
1.5615
5 g
1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane
KXROTPXCYDXGSC-UHFFFAOYSA-N
2-methyl-1,3-dithiane
22329
98%
6007-26-7
78°C to 80°C (9 mmHg)
C5H10S2
MFCD00014652
102766
Not miscible or difficult to mix in water. Soluble in alcohol.
CC1SCCCS1
134.255
134.25

Chemical Identifiers

6007-26-7
134.255
KXROTPXCYDXGSC-UHFFFAOYSA-N
22329
CC1SCCCS1
C5H10S2
MFCD00014652
1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane
2-methyl-1,3-dithiane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 227-859-6

RTECSNumber : JO5263000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only