missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazol-5-amine, 95%

Catalog No. AC432760010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432760010 1 g Glass bottle
AC432760050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432760010 Supplier Thermo Scientific Chemicals Supplier No. 432760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13382-43-9 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13382-43-9
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00226291
InChI Key GPWQHYMVUZYWIK-UHFFFAOYSA-N
Synonym 2-methylbenzo d thiazol-5-amine, 5-amino-2-methylbenzothiazole, 5-benzothiazolamine, 2-methyl, 2-methyl-5-benzothiazolamine, 2-methyl-5-aminobenzothiazole, 2-methylbenzothiazole-5-ylamine, 2-methyl-1,3-benzothiazol-5-ylamine, acmc-209v5w, cambridge id 5107842, 2-methylbenzothiazol-5-amine
PubChem CID 36229
IUPAC Name 2-methyl-1,3-benzothiazol-5-amine
SMILES CC1=NC2=CC(N)=CC=C2S1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 101°C to 103.5°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 164.23
Percent Purity 95%
Chemical Name or Material 2-Methyl-1, 3-benzothiazol-5-amine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only