missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AAH5006003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5006003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5006003 Supplier Thermo Scientific Chemicals Supplier No. H5006003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 532391-31-4
Molecular Formula C12H18BNO3
Molecular Weight (g/mol) 235.09
MDL Number MFCD05663507
InChI Key JEJYNUCGRAHMDO-UHFFFAOYSA-N
Synonym 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-methoxypyridine-3-boronic acid pinacol ester, 2-methoxypyridin-3-ylboronic acid pinacol ester, 2-methoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pubchem16545, ksc498e8t, 2-methoxypyridine-3-boronic acid, pinacol ester, 2-methoxypyridin-3-yl boronic acid pinacol ester
PubChem CID 17750166
IUPAC Name 2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES COC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 235.09
Percent Purity 97%
Chemical Name or Material 2-Methoxypyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only