missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$63.38 - $63.38
Specifications
| CAS | 163105-90-6 |
|---|---|
| Melting Point | 145°C |
| Color | White |
| Molecular Formula | C6H8BNO3 |
| MDL Number | MFCD03411572 |
Specifications
| 163105-90-6 | |
| White | |
| MFCD03411572 | |
| (2-methoxypyridin-3-yl)boronic acid | |
| 152.94 |
| 145°C | |
| C6H8BNO3 | |
| NVOLYUXUHWBCRJ-UHFFFAOYSA-N | |
| 2762709 | |
| Crystalline Powder |