missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-3-boronic acid, 98%

Catalog No. AAL2009403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2009403 1 g
AAL2009406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2009403 Supplier Thermo Scientific Chemicals Supplier No. L2009403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 163105-90-6 | C6H8BNO3 | 152.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163105-90-6
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD03411572
InChI Key NVOLYUXUHWBCRJ-UHFFFAOYSA-N
Synonym 2-methoxypyridine-3-boronic acid, 2-methoxy-3-pyridineboronic acid, 2-methoxypyridin-3-yl boronic acid, 2-methoxy-3-pyridylboronic acid, 2-methoxypyridin-3-yl-3-boronic acid, 2-methoxy-3-pyridinyl boronic acid, 2-methoxy-3-pyridyl boronic acid, 2-methoxy-3-pyridinyl-boronic acid, boronic acid, 2-methoxy-3-pyridinyl, boronic acid, b-2-methoxy-3-pyridinyl
PubChem CID 2762709
IUPAC Name (2-methoxypyridin-3-yl)boronic acid
SMILES COC1=NC=CC=C1B(O)O

Specifications

Melting Point 136°C to 138°C
Quantity 1 g
Formula Weight 152.95
Percent Purity 98%
Chemical Name or Material 2-Methoxypyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only