missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7132626
Change view
Click to view available options
Quantity:
1 g
25 g
Catalog No. Quantity
M12611G 1 g
M126125G 25 g
2 options

Catalog No. M12611G

Supplier: TCI America M12611G

Only null left

Chemical Identifiers

Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00236047
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
PubChem CID 2733958
IUPAC Name (2-methoxyphenyl)boronic acid

Specifications

CAS 5720-06-9
Melting Point 108°C
Color White-Yellow
Quantity 1 g
Synonym 2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
SMILES COC1=CC=CC=C1B(O)O
Formula Weight 151.96
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyphenylboronic Acid (contains varying amounts of Anhydride)