missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynicotinic Acid 98.0+%, TCI America™

Catalog No. p-7132987
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M20415G 5 g
M204125G 25 g
2 options

Catalog No. M20415G

Supplier: TCI America M20415G

Only null left

Chemical Identifiers

CAS 16498-81-0
Molecular Formula C7H6NO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00085110
InChI Key FTEZJSXSARPZHJ-UHFFFAOYSA-M
Synonym 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
PubChem CID 708093
IUPAC Name 2-methoxypyridine-3-carboxylate
SMILES COC1=NC=CC=C1C([O-])=O

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxynicotinic Acid