missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynicotinic acid, 98%

Catalog No. AAA1319303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1319303 1 g
AAA1319306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1319303 Supplier Thermo Scientific Chemicals Supplier No. A1319303

CAS: 16498-81-0 | C7H6NO3 | 152.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16498-81-0
Molecular Formula C7H6NO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00085110
InChI Key FTEZJSXSARPZHJ-UHFFFAOYSA-M
Synonym 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
PubChem CID 708093
IUPAC Name 2-methoxypyridine-3-carboxylic acid
SMILES COC1=NC=CC=C1C([O-])=O

Specifications

Melting Point 147°C to 151°C
Quantity 1 g
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 2-Methoxynicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only