missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethylamine 98.0+%, TCI America™

Catalog No. M061125ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
Catalog No. Quantity
M061125ML 25 mL
1 options

Catalog No. M061125ML

Supplier: TCI America M061125ML

Only null left

Chemical Identifiers

CAS 109-85-3
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00008180
InChI Key ASUDFOJKTJLAIK-UHFFFAOYSA-N
Synonym 2-methoxyethylamine, ethanamine, 2-methoxy, 2-aminoethyl methyl ether, ethylamine, 2-methoxy, 2-methoxyethan-1-amine, 1-amino-2-methoxyethane, 1-methoxy-2-aminoethane, beta-methoxyethylamine, 2-methoxy-1-ethanamine, 2-methoxy-ethylamine
PubChem CID 8018
IUPAC Name 2-methoxyethanamine
SMILES COCCN

Specifications

Boiling Point 90°C
Quantity 25 mL
UN Number 2733
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethylamine
RTECSNumber KR8750000
TSCA Yes