missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. A1405500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1405500ML 500 mL
A140525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1405500ML Supplier TCI America Supplier No. A1405500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3121-61-7
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00048149,MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 56°C
Quantity 500 mL
UN Number 1992
Formula Weight 130.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-1004
RTECSNumber KL6000000
TSCA Yes