missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol

Catalog No. p-7058379
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1190218 50 g
AAL1190230 250 g
AAL119020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1190218 Supplier Thermo Scientific Chemicals Supplier No. L1190218
Only null left

CAS: 3121-61-7 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3121-61-7
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00048149,MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Melting Point <-30°C
Density 1.008
Boiling Point 162°C
Flash Point 56°C (133°F)
Odor Ester-like
Refractive Index 1.428
Linear Formula H2C=CHCO2(CH2)2OCH3
Quantity 50 g
UN Number UN2929
Beilstein 1754333
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 2-Methoxyethyl acrylate, stabilized with ≈50 to 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H302+H332-H311-H360FD
Hazard Statement GHS H Statement
H311-H331-H226-H361-H302-H315-H319-H317
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-499-3
RTECSNumber KL6000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only