missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl Acetoacetate 95.0+%, TCI America™

Catalog No. A1583500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1583500G 500 g
A158325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1583500G Supplier TCI America Supplier No. A1583500G
Only null left

Chemical Identifiers

CAS 22502-03-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00009648
InChI Key PLHCSZRZWOWUBW-UHFFFAOYSA-N
Synonym 2-methoxyethyl acetoacetate, 2-methoxyethylacetoacetate, acetoacetic acid 2-methoxyethyl ester, butanoic acid, 3-oxo-, 2-methoxyethyl ester, methoxyethyl acetoacetate, acmc-1cs35, 2-methoxyethyl-3-oxobutanoate, #, 2-methoxy ethyl acetoacetate, 2-methoxyethyl acetoacetate, lonza quality gc
PubChem CID 89734
IUPAC Name 2-methoxyethyl 3-oxobutanoate
SMILES COCCOC(=O)CC(C)=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 500 g
Formula Weight 160.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl Acetoacetate