missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyethoxymethyl Chloride 95.0+%, TCI America™
$145.40 - $1112.84
Chemical Identifiers
| CAS | 3970-21-6 |
|---|---|
| Molecular Formula | C4H9ClO2 |
| Molecular Weight (g/mol) | 124.56 |
| MDL Number | MFCD00000888 |
| InChI Key | BIAAQBNMRITRDV-UHFFFAOYSA-N |
| Synonym | 2-methoxyethoxymethyl chloride, 1-chloromethoxy-2-methoxyethane, ethane, 1-chloromethoxy-2-methoxy, mem chloride, methoxyethoxymethyl chloride, mem-chloride, 1-chloro-2,5-dioxahexane, 2-methoxyethoxy methyl chloride, beta-methoxyethoxymethyl chloride, .beta.-methoxyethoxymethyl chloride |
| PubChem CID | 77590 |
| IUPAC Name | 1-(chloromethoxy)-2-methoxyethane |
| SMILES | COCCOCCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M068025ML
|
TCI America
M068025ML |
25 mL |
Each for $145.40
|
|
|||||
|
M0680500ML
|
TCI America
M0680500ML |
500 mL |
Each for $1,112.84
|
|
|||||
Chemical Identifiers
| 3970-21-6 | |
| 124.56 | |
| BIAAQBNMRITRDV-UHFFFAOYSA-N | |
| 77590 | |
| COCCOCCl |
| C4H9ClO2 | |
| MFCD00000888 | |
| 2-methoxyethoxymethyl chloride, 1-chloromethoxy-2-methoxyethane, ethane, 1-chloromethoxy-2-methoxy, mem chloride, methoxyethoxymethyl chloride, mem-chloride, 1-chloro-2,5-dioxahexane, 2-methoxyethoxy methyl chloride, beta-methoxyethoxymethyl chloride, .beta.-methoxyethoxymethyl chloride | |
| 1-(chloromethoxy)-2-methoxyethane |
Specifications
| 3970-21-6 | |
| 52°C | |
| MFCD00000888 | |
| 2929 | |
| BIAAQBNMRITRDV-UHFFFAOYSA-N | |
| 1-(chloromethoxy)-2-methoxyethane | |
| 77590 | |
| ≥95.0% (GC) | |
| 2-Methoxyethoxymethyl Chloride |
| Colorless | |
| C4H9ClO2 | |
| 25 mL | |
| 2-methoxyethoxymethyl chloride, 1-chloromethoxy-2-methoxyethane, ethane, 1-chloromethoxy-2-methoxy, mem chloride, methoxyethoxymethyl chloride, mem-chloride, 1-chloro-2,5-dioxahexane, 2-methoxyethoxy methyl chloride, beta-methoxyethoxymethyl chloride, .beta.-methoxyethoxymethyl chloride | |
| COCCOCCl | |
| 124.56 | |
| 124.56 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator