missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxycyclohexanone 95.0+%, TCI America™
$90.48 - $438.93
Chemical Identifiers
| CAS | 7429-44-9 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.171 |
| MDL Number | MFCD00009610 |
| InChI Key | JYJURPHZXCLFDX-UHFFFAOYSA-N |
| PubChem CID | 110998 |
| IUPAC Name | 2-methoxycyclohexan-1-one |
| SMILES | COC1CCCCC1=O |
Chemical Identifiers
| 7429-44-9 | |
| 128.171 | |
| JYJURPHZXCLFDX-UHFFFAOYSA-N | |
| 2-methoxycyclohexan-1-one |
| C7H12O2 | |
| MFCD00009610 | |
| 110998 | |
| COC1CCCCC1=O |
Specifications
| 7429-44-9 | |
| 88°C | |
| MFCD00009610 | |
| JYJURPHZXCLFDX-UHFFFAOYSA-N | |
| 2-methoxycyclohexan-1-one | |
| 110998 | |
| ≥95.0% (GC) | |
| 2-Methoxycyclohexanone |
| Yellow | |
| C7H12O2 | |
| 5 g | |
| COC1CCCCC1=O | |
| 128.171 | |
| 128.17 | |
| Liquid |
Safety and Handling
TSCA : Yes