missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxycyclohexanone 95.0+%, TCI America™

Catalog No. M11715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11715G 5 g
M117125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11715G Supplier TCI America Supplier No. M11715G
Only null left

Chemical Identifiers

CAS 7429-44-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009610
InChI Key JYJURPHZXCLFDX-UHFFFAOYSA-N
PubChem CID 110998
IUPAC Name 2-methoxycyclohexan-1-one
SMILES COC1CCCCC1=O

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 128.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxycyclohexanone