missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$59.69 - $59.69
Chemical Identifiers
| CAS | 374538-03-1 |
|---|---|
| Molecular Formula | C8H9BO4 |
| Molecular Weight (g/mol) | 179.97 |
| MDL Number | MFCD02179452 |
| InChI Key | ODAXNYMENLFYMY-UHFFFAOYSA-N |
| PubChem CID | 2773496 |
| IUPAC Name | [2-(methoxycarbonyl)phenyl]boronic acid |
| SMILES | COC(=O)C1=CC=CC=C1B(O)O |
Chemical Identifiers
| 374538-03-1 | |
| 179.97 | |
| ODAXNYMENLFYMY-UHFFFAOYSA-N | |
| [2-(methoxycarbonyl)phenyl]boronic acid |
| C8H9BO4 | |
| MFCD02179452 | |
| 2773496 | |
| COC(=O)C1=CC=CC=C1B(O)O |
Specifications
| 374538-03-1 | |
| C8H9BO4 | |
| 1 g | |
| ODAXNYMENLFYMY-UHFFFAOYSA-N | |
| [2-(methoxycarbonyl)phenyl]boronic acid | |
| 2773496 | |
| Crystalline Powder |
| 104°C | |
| MFCD02179452 | |
| 2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid | |
| COC(=O)C1=CC=CC=C1B(O)O | |
| 179.97 | |
| 179.97 | |
| 2-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No