missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M19051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19051G Supplier TCI America Supplier No. M19051G

Chemical Identifiers

CAS 374538-03-1
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02179452
InChI Key ODAXNYMENLFYMY-UHFFFAOYSA-N
PubChem CID 2773496
IUPAC Name [2-(methoxycarbonyl)phenyl]boronic acid
SMILES COC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 104°C
Quantity 1 g
Synonym 2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
Formula Weight 179.97
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)